Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tox2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001098799.1 | NP_001092269.1 | 3 | 237 | ||
GenBank mRNA | BB649411.1 | - | 3 | 237 | ||
BF470404.1 | - | 1 | 33 |