Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Chd5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081376.1 | NP_001074845.1 | 28 | 4495 | ||
NM_029216.2 | NP_083492.2 | 27 | 4384 | |||
GenBank mRNA | AK129145.1 | - | 4 | 464 | ||
AK147563.1 | - | 27 | 4385 |