Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Gcdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001044744.1 | NP_001038209.2 | 10 | 1194 | ||
NM_008097.2 | NP_032123.3 | 9 | 1251 | |||
GenBank mRNA | AK078866.1 | - | 7 | 804 | ||
AK079500.1 | - | 3 | 302 | |||
AK165406.1 | - | 9 | 1077 | |||
BC061158.1 | - | 9 | 1081 | 30314273 | ||
U18992.1 | - | 9 | 1032 |