Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Lrrc36 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033371.3 | NP_001028543.1 | 6 | 1075 | ||
NM_001170788.1 | NP_001164259.1 | 9 | 1404 | |||
NM_001170789.1 | NP_001164260.1 | 6 | 1081 | |||
GenBank mRNA | AK133042.1 | - | 6 | 1075 | ||
BC036564.1 | - | 4 | 703 | |||
BC150957.1 | - | 6 | 1067 | 9087868 | ||
BC172120.1 | - | 6 | 1073 | 9054406 |