Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Snf8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_033568.2 | NP_291046.1 | 8 | 843 | ||
GenBank mRNA | AK013058.1 | - | 8 | 845 | ||
AK020203.1 | - | 6 | 533 | |||
AK180940.1 | - | 2 | 153 | |||
BC003938.1 | - | 8 | 815 | 3486703 | ||
BC030490.1 | - | 2 | 1648 |