Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nynrin | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001040072.1 | NP_001035161.1 | 3 | 1858 | ||
GenBank mRNA | BC151043.1 | - | 4 | 1880 | 9087956 |