Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Miip | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001025365.2 | NP_001020536.2 | 5 | 1018 | ||
GenBank mRNA | AK133976.1 | - | 7 | 1170 | ||
AK170224.1 | - | 5 | 811 | |||
BC017525.1 | - | 5 | 761 | 4910951 | ||
BC024069.1 | - | 5 | 800 | 5097088 | ||
FJ618908.1 | - | 5 | 1018 |