Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Igfbp7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001159518.1 | NP_001152990.1 | 4 | 753 | ||
NM_008048.3 | NP_032074.3 | 3 | 660 | |||
GenBank mRNA | AA200937.1 | - | 3 | 456 | ||
AB012886.1 | - | 3 | 661 | |||
AK002277.1 | - | 3 | 656 | |||
AK148749.1 | - | 3 | 657 | |||
AK156315.1 | - | 3 | 662 | |||
BC047202.1 | - | 3 | 572 | |||
BC092538.1 | - | 3 | 656 | 30284911 | ||
BF471478.1 | - | 4 | 267 | |||
CK622921.1 | - | 3 | 463 | |||
L75822.2 | - | 3 | 639 |