Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Apln | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_013912.3 | NP_038940.1 | 2 | 474 | ||
GenBank mRNA | AB023494.1 | - | 2 | 127 | ||
AJ290423.1 | - | 2 | 226 | |||
AK042076.1 | - | 2 | 478 | |||
BB618863.1 | - | 2 | 474 | |||
BC020015.1 | - | 2 | 455 | 3483588 | ||
DQ832282.1 | - | 2 | 463 |