Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Celf5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176954.3 | NP_795928.2 | 3 | 357 | ||
GenBank mRNA | AK029796.1 | - | 3 | 1534 | ||
AK083461.1 | - | 2 | 85 | |||
AK132321.1 | - | 3 | 323 | |||
AK158669.1 | - | 2 | 134 | |||
BC079845.1 | - | 3 | 332 |