Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Msrb3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177092.4 | NP_796066.1 | 4 | 398 | ||
GenBank mRNA | AK086975.1 | - | 4 | 365 | ||
AK137163.1 | - | 4 | 365 | |||
BB617432.1 | - | 4 | 433 | |||
BC096447.1 | - | 4 | 279 | 6310686 |