Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Heatr5a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177171.4 | NP_796145.2 | 36 | 6106 | ||
GenBank mRNA | AK122493.1 | - | 25 | 4110 | ||
AK161444.1 | - | 25 | 4425 | |||
AK161523.1 | - | 8 | 1466 | |||
BC063265.1 | - | 12 | 2119 | |||
BC086656.1 | - | 35 | 5931 | 30544145 |