Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tmem88b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033394.3 | NP_001028566.2 | 1 | 375 | ||
GenBank mRNA | AK035998.1 | - | 1 | 375 | ||
AK158459.1 | - | 1 | 376 | |||
BC147318.1 | - | 1 | 330 | 9056362 | ||
BC147319.1 | - | 1 | 330 | 9056365 |