Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Samd12 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177225.4 | NP_796199.1 | 1, 2 | 74 | ||
GenBank mRNA | AK044142.1 | - | 1, 2 | 75 | ||
BC119383.1 | - | 1, 2 | 75 | 8734132 | ||
BC119385.1 | - | 1, 2 | 75 | 8734134 | ||
BC144982.1 | - | 1, 2 | 75 | 9053530 |