Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pla2g4e | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177845.4 | NP_808513.2 | 17 | 2445 | ||
GenBank mRNA | AB195277.1 | - | 17 | 2348 | ||
BC060600.1 | - | 14 | 2391 | |||
BC066085.1 | - | 17 | 2445 | |||
BC137899.1 | - | 17 | 2222 | 8860918 | ||
BC145020.1 | - | 18 | 2333 |