Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fnip2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001162999.2 | NP_001156471.1 | 13 | 2688 | ||
GenBank mRNA | AK122512.1 | - | 5 | 1765 | ||
AK141700.1 | - | 1 | 809 | |||
BC116316.1 | - | 6 | 1952 | |||
BC116317.1 | - | 6 | 1952 |