Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Wipf3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001167860.1 | NP_001161332.1 | 7, 8 | 1426 | ||
NM_001167861.1 | NP_001161333.1 | 6, 7 | 1324 |