Search Thermo Fisher Scientific
- Contact Us
- Quick Order
-
Don't have an account ? Create Account
Search Thermo Fisher Scientific
Gene Symbol: Ccdc27 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001033455.2 | NP_001028627.1 | 1 | 141 | ||
GenBank mRNA | AK133459.1 | - | 1 | 142 | ||
BC150930.1 | - | 1 | 124 | 9087841 | ||
BC151167.1 | - | 1 | 124 | 9088056 |