Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kpna7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001013774.2 | NP_001013796.2 | 7, 8 | 866 | ||
GenBank mRNA | AK136027.1 | - | 3, 4 | 384 | ||
AY950703.1 | - | 7, 8 | 867 | |||
FJ717332.1 | - | 5, 6 | 633 | |||
FJ717333.1 | - | 6, 7 | 696 |