Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ces3a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001164681.1 | NP_001158153.1 | 9 | 1118 | ||
NM_198672.1 | NP_941074.1 | 10 | 1259 | |||
GenBank mRNA | AK138932.1 | - | 10 | 1258 | ||
AK149499.1 | - | 6 | 679 | |||
BC035526.1 | - | 9 | 1114 | |||
BC048380.1 | - | 10 | 1240 | |||
BC057187.1 | - | 10 | 1244 | 5098677 | ||
BC057188.1 | - | 10 | 1244 | 5100121 | ||
BC061004.1 | - | 9 | 1116 | 30315941 | ||
L11333.1 | - | 10 | 1234 | |||
S64130.1 | - | 10 | 1235 |