Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pnmal2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001099636.2 | NP_001093106.1 | 1 | 1226 | ||
GenBank mRNA | AK140282.1 | - | 1 | 1227 | ||
AK143183.1 | - | 1 | 1205 |