Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Hist1h1c | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015786.3 | NP_056601.1 | 1 | 555 | ||
GenBank mRNA | AK002321.1 | - | 555 | |||
AK135614.1 | - | 287 | ||||
AK155502.1 | - | 556 | ||||
AK209931.1 | - | 1 | 153 | |||
AK211271.1 | - | 1 | 65 | |||
BC089604.1 | - | 555 | 6763721 |