Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fbxl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_015822.2 | NP_056637.1 | 2 | 571 | ||
GenBank mRNA | AF176521.1 | - | 2 | 278 | ||
AK080863.1 | - | 2 | 580 | |||
AK142007.1 | - | 2 | 571 | |||
AK166128.1 | - | 2 | 452 | |||
AK167409.1 | - | 2 | 390 | |||
BC026835.1 | - | 1 | 237 | |||
BC037665.1 | - | 1 | 234 | |||
BC067203.1 | - | 2 | 453 | 6334317 |