Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Preb | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001294302.1 | NP_001281231.1 | 1 | 270 | ||
NM_016703.3 | NP_057912.2 | 1 | 270 | |||
GenBank mRNA | AF150808.1 | - | 1 | 173 | ||
AF193017.1 | - | 1 | 185 | |||
AK013330.1 | - | 1 | 224 | |||
AK029838.1 | - | 1 | 205 | |||
AK036866.1 | - | 1 | 206 | |||
AK151288.1 | - | 1 | 252 | |||
BB846175.1 | - | 1 | 305 | |||
BC017527.1 | - | 1 | 173 | 4921119 |