Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pvr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027514.2 | NP_081790.1 | 2 | 438 | ||
GenBank mRNA | AB195430.1 | - | 2 | 395 | ||
AJ563420.1 | - | 2 | 326 | |||
AK014446.1 | - | 2 | 409 | |||
AK038747.1 | - | 2 | 409 | |||
AK040334.1 | - | 2 | 409 | |||
AK078344.1 | - | 2 | 405 | |||
AK080753.1 | - | 2 | 409 | |||
AK165873.1 | - | 2 | 428 | |||
BC013673.1 | - | 2 | 395 | 4188634 | ||
BC032283.1 | - | 2 | 398 | 5363187 | ||
BC138219.1 | - | 2 | 383 | 8861238 | ||
BC138220.1 | - | 2 | 383 | 8861239 | ||
BY276828.1 | - | 2 | 439 |