Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pvr | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_027514.2 | NP_081790.1 | 3 | 603 | ||
GenBank mRNA | AB195430.1 | - | 3 | 560 | ||
AJ563420.1 | - | 3 | 491 | |||
AK014446.1 | - | 3 | 574 | |||
AK038747.1 | - | 3 | 574 | |||
AK078344.1 | - | 3 | 570 | |||
AK080753.1 | - | 3 | 574 | |||
AK165873.1 | - | 3 | 593 | |||
BC013673.1 | - | 3 | 560 | 4188634 | ||
BC032283.1 | - | 3 | 563 | 5363187 | ||
BC138219.1 | - | 3 | 548 | 8861238 | ||
BC138220.1 | - | 3 | 548 | 8861239 | ||
U35836.1 | - | 2 | 522 |