Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cbx7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_144811.3 | NP_659060.1 | 5, 6 | 456 | ||
GenBank mRNA | AB221539.1 | - | 5, 6 | 330 | ||
AK045714.1 | - | 5, 6 | 730 | |||
AY453793.1 | - | 5, 6 | 446 | |||
BC021398.1 | - | 5, 6 | 472 | 5066655 | ||
BC030919.1 | - | 1, 2 | 323 | |||
BQ571764.1 | - | 3, 4 | 223 |