Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fut8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001252614.1 | NP_001239543.1 | 13 | 2191 | ||
NM_001252615.1 | NP_001239544.1 | 7 | 1265 | |||
NM_001252616.1 | NP_001239545.1 | 6 | 1285 | |||
NM_016893.5 | NP_058589.2 | 12 | 2102 | |||
GenBank mRNA | AB025198.1 | - | 9 | 1791 | ||
AK048520.1 | - | 12 | 2240 | |||
AK051811.1 | - | 12 | 2059 | |||
AK134537.1 | - | 12 | 2195 | |||
AK183245.1 | - | 2 | 198 | |||
BC010666.1 | - | 13 | 2170 | 3594582 | ||
EF591874.1 | - | 7 | 1265 | |||
EF591875.1 | - | 6 | 1285 |