Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Caprin1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001111289.1 | NP_001104759.1 | 5 | 754 | ||
NM_001111290.1 | NP_001104760.1 | 5 | 754 | |||
NM_001111291.1 | NP_001104761.1 | 6 | 794 | |||
NM_001111292.1 | NP_001104762.1 | 5 | 754 | |||
NM_016739.3 | NP_058019.2 | 6 | 794 | |||
GenBank mRNA | AB373955.1 | - | 5 | 616 | ||
AK133891.1 | - | 6 | 756 | |||
AK141992.1 | - | 5 | 696 | |||
AK147514.1 | - | 6 | 761 | |||
AK148203.1 | - | 5 | 741 | |||
AK157707.1 | - | 6 | 778 | |||
AK166461.1 | - | 5 | 712 | |||
AK168676.1 | - | 5 | 749 | |||
AK170830.1 | - | 6 | 756 | |||
AK214887.1 | - | 2 | 229 | |||
AK219211.1 | - | 2 | 229 | |||
BC052427.1 | - | 6 | 773 | 6836041 | ||
U27838.1 | - | 5 | 731 |