Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Celsr2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001004177.2 | NP_001004177.2 | 4 | 4200 | ||
NM_017392.3 | NP_059088.2 | 4 | 4200 | |||
GenBank mRNA | AB028499.1 | - | 4 | 4203 | ||
AK220331.1 | - | 3 | 587 | |||
CD350951.1 | - | 2 | 252 |
Set Membership: |
G-protein coupled receptor |