Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fam69b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019833.3 | NP_062807.2 | 4 | 515 | ||
GenBank mRNA | AB030186.1 | - | 4 | 510 | ||
AF332189.1 | - | 4 | 509 | |||
AK045880.1 | - | 4 | 516 | |||
AK050584.1 | - | 4 | 429 | |||
AK171112.1 | - | 3 | 4375 | |||
BC051954.1 | - | 3 | 287 | 5714977 | ||
BC060081.1 | - | 3 | 287 | 5714977 | ||
BC116751.1 | - | 4 | 302 | 40126070 | ||
BC116779.1 | - | 4 | 302 |