Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fzr1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019757.1 | NP_062731.1 | 5 | 451 | ||
GenBank mRNA | AF083809.1 | - | 5 | 451 | ||
AK051993.1 | - | 5 | 533 | |||
AK081476.1 | - | 5 | 505 | |||
AK154822.1 | - | 5 | 535 | |||
AK155370.1 | - | 5 | 533 | |||
AK163951.1 | - | 5 | 510 | |||
AK172679.1 | - | 5 | 545 | |||
BC006616.1 | - | 5 | 532 | 3967009 |