Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Morf4l2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001168225.1 | NP_001161697.1 | 4 | 1090 | ||
NM_001168226.1 | NP_001161698.1 | 4 | 1023 | |||
NM_001168227.1 | NP_001161699.1 | 4 | 975 | |||
NM_001168228.1 | NP_001161700.1 | 4 | 974 | |||
NM_001168229.1 | NP_001161701.1 | 3 | 946 | |||
NM_001168230.1 | NP_001161702.1 | 3 | 884 | |||
NM_019768.4 | NP_062742.4 | 4 | 1097 | |||
GenBank mRNA | AB025049.1 | - | 2 | 785 | ||
AK075946.1 | - | 4 | 950 | |||
AK088849.1 | - | 1 | 804 | |||
AK089963.1 | - | 4 | 951 | |||
AK129038.1 | - | 4 | 931 | |||
AK131944.1 | - | 4 | 950 | |||
AK145686.1 | - | 4 | 950 | |||
AK150401.1 | - | 4 | 950 | |||
AK150670.1 | - | 4 | 950 | |||
AK151570.1 | - | 4 | 950 | |||
AK153146.1 | - | 4 | 946 | |||
AK159145.1 | - | 4 | 999 | |||
AK159179.1 | - | 4 | 950 | |||
AK160012.1 | - | 4 | 950 | |||
AK161610.1 | - | 4 | 976 | |||
AK164433.1 | - | 4 | 950 | |||
AK165985.1 | - | 4 | 950 | |||
AK166637.1 | - | 4 | 976 | |||
AK192830.1 | - | 1 | 188 | |||
BC075653.1 | - | 4 | 1097 | 30539330 | ||
BC088731.1 | - | 1 | 731 | 5340043 |