Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Srpk3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019684.1 | NP_062658.1 | 2 | 115 | ||
GenBank mRNA | AF043288.1 | - | 2 | 115 | ||
AK004696.2 | - | 2 | 133 | |||
BC120883.1 | - | 2 | 110 | 40057934 | ||
BC120884.1 | - | 2 | 110 | 40057937 |