Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pf4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_019932.4 | NP_064316.1 | 2 | 261 | ||
GenBank mRNA | AB017491.2 | - | 2 | 196 | ||
AK159278.1 | - | 2 | 261 | |||
AK160040.1 | - | 2 | 261 | |||
BC061111.1 | - | 2 | 193 | 30280161 |