Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Ascl3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020051.1 | NP_064435.1 | 2 | 467 | ||
GenBank mRNA | AB046448.1 | - | 2 | 453 | ||
AB221543.1 | - | 1 | 407 | |||
AJ277605.1 | - | 2 | 437 | |||
BC104346.1 | - | 2 | 462 | 40049496 | ||
BC104347.1 | - | 2 | 461 | 40049502 |