Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tssc4 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001115085.1 | NP_001108557.1 | 3 | 510 | ||
NM_020285.2 | NP_064681.1 | 3 | 540 | |||
NM_138631.1 | NP_619537.1 | 3 | 540 | |||
GenBank mRNA | AB038011.1 | - | 3 | 540 | ||
AB041597.1 | - | 1 | 881 | |||
AJ279796.1 | - | 3 | 540 | |||
AJ279797.1 | - | 3 | 540 | |||
AK088052.1 | - | 3 | 475 | |||
BC131974.1 | - | 2 | 352 | |||
BC131976.1 | - | 2 | 352 | |||
BC145365.1 | - | 2 | 352 | |||
CX239537.1 | - | 3 | 454 |