Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pdxp | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_020271.3 | NP_064667.2 | 2 | 764 | ||
GenBank mRNA | AB041662.1 | - | 2 | 659 | ||
AK043228.1 | - | 2 | 725 | |||
AY366300.1 | - | 2 | 726 | |||
BC037010.1 | - | 2 | 189 | |||
BC046955.1 | - | 2 | 715 | |||
BC049107.1 | - | 2 | 764 | |||
BC058388.1 | - | 2 | 310 | |||
BC069982.1 | - | 2 | 536 | |||
BU148105.1 | - | 2 | 533 |