Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Psmd8 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_026545.3 | NP_080821.3 | 1 | 319 | ||
GenBank mRNA | AK002921.1 | - | 1 | 158 | ||
AK003436.1 | - | 1 | 160 | |||
AK020132.1 | - | 1 | 178 | |||
AK027973.1 | - | 1 | 123 | |||
AK051257.1 | - | 1 | 171 | |||
AK150493.1 | - | 1 | 162 | |||
AK150982.1 | - | 1 | 126 | |||
AK153329.1 | - | 1 | 160 | |||
AK159216.1 | - | 1 | 128 | |||
AK159795.1 | - | 1 | 144 | |||
AK159926.1 | - | 1 | 154 | |||
AK166718.1 | - | 1 | 153 | |||
AK167948.1 | - | 1 | 128 | |||
AK168886.1 | - | 1 | 127 | |||
BC004075.1 | - | 1 | 137 | 3591796 | ||
BC005717.1 | - | 1 | 47 | 2609781 | ||
CF732286.1 | - | 1 | 319 |