Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fxyd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022007.1 | NP_071290.1 | 3 | 163 | ||
GenBank mRNA | AI892161.1 | - | 3 | 163 | ||
AK004113.1 | - | 3 | 172 | |||
BC048737.1 | - | 3 | 176 | |||
BC061101.1 | - | 3 | 176 | 6531288 |