Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fxyd7 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022007.1 | NP_071290.1 | 1, 2 | 102 | ||
GenBank mRNA | AI892161.1 | - | 1, 2 | 102 | ||
AK004113.1 | - | 1, 2 | 111 | |||
BC048737.1 | - | 1, 2 | 115 | |||
BC061101.1 | - | 1, 2 | 115 | 6531288 |