Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Pidd1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_022654.1 | NP_073145.1 | 3 | 925 | ||
GenBank mRNA | AF274973.1 | - | 3 | 925 | ||
AK004702.1 | - | 4 | 926 | |||
AK156863.1 | - | 5 | 913 | |||
BC032202.1 | - | 3 | 944 | |||
BC056987.1 | - | 3 | 1276 | |||
BC089373.1 | - | 3 | 897 | |||
BC145314.1 | - | 3 | 929 | |||
BC145315.1 | - | 3 | 890 | |||
BC145857.1 | - | 3 | 890 | 40131160 |