Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Cacna1h | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163691.1 | NP_001157163.1 | 6 | 1025 | ||
NM_021415.4 | NP_067390.4 | 6 | 1025 | |||
GenBank mRNA | AK147242.1 | - | 6 | 1026 | ||
BC058206.1 | - | 6 | 1010 | |||
BC138026.1 | - | 6 | 864 | 8861045 |