Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Nap1l5 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021432.2 | NP_067407.1 | 1 | 351 | ||
GenBank mRNA | AB041556.1 | - | 1 | 290 | ||
AK003875.1 | - | 1 | 57 | |||
BC046332.1 | - | 1 | 301 | 6491916 | ||
BU526293.1 | - | 1 | 354 |