Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Tdh | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021480.5 | NP_067455.5 | 4, 5 | 696 | ||
GenBank mRNA | AF134346.2 | - | 4, 5 | 360 | ||
AK077571.1 | - | 4, 5 | 371 | |||
AY116662.1 | - | 3, 4 | 297 | |||
BC058860.1 | - | 4, 5 | 711 | 30014660 |