Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Rps6kb2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021485.2 | NP_067460.1 | 12 | 996 | ||
GenBank mRNA | AJ007938.1 | - | 12 | 996 | ||
AK014412.1 | - | 12 | 1075 | |||
AK090093.1 | - | 1051 | ||||
AK156361.1 | - | 12 | 1050 | |||
BC005694.1 | - | 4 | 327 | |||
BC051631.1 | - | 12 | 1302 | |||
BC066842.1 | - | 11 | 1413 | 30468251 |