Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Thap11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021513.2 | NP_067488.1 | 1 | 1034 | ||
GenBank mRNA | AB041579.1 | - | 1 | 995 | ||
AK165981.1 | - | 1 | 995 | |||
AK166109.1 | - | 1 | 1031 | |||
AK170457.1 | - | 1 | 1006 | |||
BC003949.1 | - | 1 | 587 | |||
BC138966.1 | - | 1 | 946 | 8861988 | ||
BC138987.1 | - | 1 | 946 | 8862009 |