Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Fancg | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001163233.1 | NP_001156705.1 | 3 | 431 | ||
NM_053081.2 | NP_444311.1 | 3 | 431 | |||
GenBank mRNA | AF112439.1 | - | 3 | 240 | ||
AY049715.1 | - | 3 | 231 | |||
BC050890.1 | - | 4 | 535 | |||
BC139763.1 | - | 3 | 234 | 40110334 | ||
BC141282.1 | - | 3 | 247 | 9055894 |