Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Foxj2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021899.3 | NP_068699.1 | 3 | 1149 | ||
GenBank mRNA | AB221666.1 | - | 2 | 348 | ||
AF253052.1 | - | 3 | 1161 | |||
AK154574.1 | - | 3 | 1392 | |||
BC040395.1 | - | 4 | 779 | 4190087 |