Search Thermo Fisher Scientific
Search Thermo Fisher Scientific
Gene Symbol: Kctd21 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039039.3 | NP_001034128.2 | 2 | 815 | ||
GenBank mRNA | AK141547.1 | - | 2 | 812 | ||
BC139363.1 | - | 1 | 764 | 8862385 | ||
BC139365.1 | - | 1 | 764 | 8862387 | ||
CX221410.1 | - | 1 | 276 |